Anti-NT3 Sheep Polyclonal Antibody

Leverandør: Biosensis
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSENS-060-50EA 2260 DKK
BSENS-060-50
Anti-NT3 Sheep Polyclonal Antibody
Antistoffer
FUNCTION: Seems to promotes the survival of visceral and proprioceptive sensory neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family.

A cross reactivity of less than 0.1% to mouse NGF, recombinant human BDNF and NT4/5 has been shown by dot blot.
Application Information:
IHC, ELISA, WB, inhibition of biological activity. A concentration of 1-10 µg/ml is recommended for IHC, ELISA, WB and inhibition of biological activity in vitro; 2-10 µg/ml (ED50) for in vivo use. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: central region of NT3
Host: Sheep
Isotype: IgG
Reactivity: Rat, Chicken, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR