Anti-TLE1 Mouse monoclonal antibody [clone: 1F5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131648-100EA 0 DKK
ABCAAB131648-100
Anti-TLE1 Mouse monoclonal antibody [clone: 1F5]
Antistoffer
Type: Primary
Antigen: TLE1 (transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila))
Clonality: monoclonal
Clone: 1F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR