Anti-GP1BA Mouse Monoclonal Antibody (CF®405M) [clone: HIP1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB119496-50EA 0 DKK
ABCAAB119496-50
Anti-GP1BA Mouse Monoclonal Antibody (CF®405M) [clone: HIP1]
Antistoffer
Type: Primary
Antigen: GP1BA (glycoprotein Ib (platelet), alpha polypeptide)
Clonality: monoclonal
Clone: HIP1
Conjugation: CF® 405M Dye
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR