Anti-HCN3 Mouse monoclonal antibody [clone: S141-28]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB84818-100EA 0 DKK
ABCAAB84818-100
Anti-HCN3 Mouse monoclonal antibody [clone: S141-28]
Antistoffer
Type: Primary
Antigen: HCN3 (hyperpolarization activated cyclic nucleotide-gated potassium channel 3)
Clonality: monoclonal
Clone: S141-28
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR