Anti-NCK1 Mouse monoclonal antibody [clone: 11D10]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB77124-100EA 0 DKK
ABCAAB77124-100
Anti-NCK1 Mouse monoclonal antibody [clone: 11D10]
Antistoffer
Type: Primary
Antigen: NCK1 (NCK adaptor protein 1)
Clonality: monoclonal
Clone: 11D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR