Anti-ARRB1 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB31868-100EA 0 DKK
ABCAAB31868-100
Anti-ARRB1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: ARRB1 (arrestin, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR