Gram Negative lysis buffer
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-566 786-567 786-579 786-580
786-566EA 805 DKK
786-566 786-567 786-579 786-580
Gram Negative lysis buffer
Buffere Ekstraktions buffere
Gram-negative lysis buffer is an extraction buffer for soluble proteins from Gram-negative bacteria. It is a proprietary improvement on the lysozyme based lysis in combination with various salts and agents, which allows extraction of soluble proteins. This buffer has been tested for use with several widely used bacteria including E.coli strains.