VWR® SuperClear, Microcentrifuge Tubes with Screw Caps
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
525-0540EA 1110 DKK
525-0540 525-0538 525-0548 525-0547 525-0546
VWR® SuperClear, Microcentrifuge Tubes with Screw Caps
Rør Mikrocentrifugerør
Resin, ultra-clear with elastomeric sealing screw caps.
  • Elastomeric integral moulded cap
  • 0,5/1,5 and 2 ml sizes
  • Reseable bags
  • Sustainable design based on virgin resins
SuperClear screw cap microtubes are made from an ultra-clear resin that gives you an excellent view of tube contents. These tubes include universal screw threads that will work with all major brands of caps. Included are large external graduations and a smooth surface for applying labels. Caps and tubes are autoclavable and freezable. Sterile tubes are radiation sterilised. Tubes can be centrifuged to 20000×g.
Caps included a moulded in place elastomeric seal to insure a leakproof seal.