Standard tube holders, VACUETTE®
Leverandør: Greiner bio-one
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VACUETTE®
GREI450209_100EA 36.4 DKK
GREI450209_100 GREI450201_1200
Standard tube holders, VACUETTE®
Kanyleholder
These VACUETTE® standard tube holders are non-sterile and made of plastic.
  • Special, user-friendly design
  • Good grip when drawing blood due to structured surface
  • Safer collection process
They are compatible with all VACUETTE® needles, VACUETTE® Luer™ adapters and blood collection sets. The use of these products with needles from other manufacturers is the responsibility of the user.