HPLC columns, ProPac™ IMAC-10
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HYPE063272EA 3940 DKK
HYPE063272 HYPE063617 HYPE063280 HYPE063615 HYPE063278 HYPE063276 HYPE063274
HPLC columns, ProPac™ IMAC-10
These columns are used to separate the metal binding proteins and peptides by immobilised metal affinity chromatography.
  • Resolve target protein
  • High loading capacity for protein purification separations
  • Wide range of metal specific applications