SRK skumspatler
Leverandør: COPAN Diagnostics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SRK®
612-2680EA 938 DKK
612-2680 612-2679 612-2678
SRK skumspatler
Podepinde Podepinde med renseopløsning
Swab rinse kits (SRK®) for microbiological examination of surfaces.
  • Substances to neutralise and deactivate disinfectants and sanitising agents
  • Moisten the sterile swab before sampling and transport the swab after collection
  • Small spatula attached to cap or large foam spatula (22 cm long (612-2678))
Can be used for microbial detection or quantitative analysis.
Emballering: In a plastic peel pouch.