Anti-CD66b Mouse Monoclonal Antibody [clone: G10F5] (PE)
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60086PE 60086PE.1
STMC60086PEEA 3377 DKK
STMC60086PE STMC60086PE.1
Anti-CD66b Mouse Monoclonal Antibody [clone: G10F5] (PE)
Antistoffer
Mouse monoclonal IgM antibody against human, chimpanzee CD66b, PE­conjugated.
  • Granulocytes
  • Subsets
  • Myeloid cells
The G10F5 antibody reacts with CD66b (previously denoted CD67), an ~95 kDa single-chain glycosylphosphatidylinositol (GPI)-linked glycoprotein expressed by neutrophilic and eosinophilic granulocytes. CD66b is a member of the carcinoembryonic antigen (CEA)-like subfamily of proteins within the immunoglobulin superfamily and possesses two Ig-like C2-type domains and one Ig-like V-type domain. In neutrophils, CD66b is localized within granulesMore Product Information