Anti-PAX6 (AN) Rabbit Polyclonal Antibody [clone: Poly] ()
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100-1340
STMC100-1340EA 6002 DKK
STMC100-1340
Anti-PAX6 (AN) Rabbit Polyclonal Antibody [clone: Poly] ()
Antistoffer
Rabbit polyclonal antibody against human, mouse PAX6 (AN).
  • Neural cells Psc derived
  • Neural stem
  • Progenitor cells
This rabbit polyclonal antibody reacts with human and mouse paired box protein Pax6, a ~46 kDa protein encoded by PAX6 gene belonging to the PAX family of transcription factors. Use this polyclonal antibody in applications requiring detection of PAX-6, such as ELISA, immunohistochemistry (IHC), western blotting (WB), immunocytochemistry (ICC), and immunofluorescence (IF). This antibody clone has been verified for labeling PAX6-positive hPSC-More Product Information