Human Recombinant Osteopontin, His tag
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100-1296
STMC100-1296EA 2960 DKK
STMC100-1296
Human Recombinant Osteopontin, His tag
Proteiner
A member of the SIBLING family of glycoproteins (Rangaswami et al.), Osteopontin (OPN) functions as T-helper 1 cytokine, and plays an important role in cell signaling, migration, and activation (Weber et al., 2002). Osteopontin has been shown to regulate inflammation both in vitro and in vivo (Agnholt et al.; Kiefer et al.), and has been implicated in the pathophysiology of numerous cancers, including prostate (Thalmann et al.), lung (Chambers et al.), ovarian (Kim et al.), and breast etiologies (Weber et al., 2015). In humans, OPN is initially secreted as a 317 amino acid protein which is subject to tissue-specific post-translational modifications by glycosylation, phosphorylation, and transglutamination (Kazanecki et al.; Sodek et al.). This protein product contains a His-residue tag at the amino end of the polypeptide chain. For consistency and reproducibility across your applications, Osteopontin from STEMCELL comes lyophilised with ≥92% purity, and endotoxin levels are verified to be ≤1,0 EU/μg protein.
Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').