RoboSep™-C Human CD4+ T Cell Isolation Kit
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100-0200
STMC100-0200EA 0 DKK
STMC100-0200
RoboSep™-C Human CD4+ T Cell Isolation Kit
Celle separations medie
Immunomagnetic negative isolation of untouched CD4+ T cells from leukopaks.
  • Fully automated large-scale isolation in a closed system
  • Up to 98% purity with high recovery
  • Untouched, viable cells
The RoboSep™-C Human CD4+ T Cell Isolation Kit is designed to isolate untouched and highly purified CD4+ T cells from fresh human peripheral blood leukopaks using RoboSep™-C (STMC100-0185 and #100-0186) for automated immunomagnetic negative selection.
This kit targets non-CD4+ T cells for removal with antibodies recognising specific cell surface markers.More Product Information