Anti-IDH1 Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5819
PRSI5819EA 3250 DKK
PRSI5819
Anti-IDH1 Rabbit Polyclonal Antibody
Antistoffer
IDH1 Antibody: Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Two NADP(+)-dependent isocitrate dehydrogenases have been found as homodimer: IDH1 is predominantly cytosolic and peroxisomal and IDH2 is mitochondrial. The presence of IDH1 in peroxisomes suggests it may play a role in the regeneration of NADPH for intraperoxisomal reductions, such as the conversion of 2, 4-dienoyl-CoAs to 3-enoyl-CoAs, as well as inMore Product Information