Anti-UROC1 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

55-667
PRSI55-667EA 4630 DKK
PRSI55-667
Anti-UROC1 Rabbit Polyclonal Antibody
Antistoffer
This gene encodes an enzyme involved in histidine catabolism, metabolizing urocanic acid to formiminoglutamic acid. The gene product is known to protect the skin from ultra violet rays and is contained in human sweat. Deficiency of this gene product in the liver is an apparent cause of mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene.

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.

Type: Primary
Antigen: UROC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR