Anti-MAZ Rabbit Polyclonal Antibody
Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25-411
PRSI25-411EA 4930 DKK
PRSI25-411
Anti-MAZ Rabbit Polyclonal Antibody
Antistoffer
MAZ may function as a transcription factor with dual roles in transcription initiation and termination.It binds to two sites, ME1a1 and ME1a2, within the c-myc promoter having greater affinity for the former. It also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.