Denaturing lysis buffer

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

J60132.AK
J60132.AKEA 476 DKK
J60132.AK
Denaturing lysis buffer
Buffere Ekstraktions buffere

Physical Form: Liquid
Appearance: Clear liquid
pH: 7,4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR