Mutagenesis kits QuikChange
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
200513 200514 200515 200516 200517 200518 200519 200531
AGLS200513EA 24515 DKK
AGLS200513 AGLS200514 AGLS200515 AGLS200516 AGLS200517 AGLS200518 AGLS200519 AGLS200531
Mutagenesis kits QuikChange
Nukleinsyrereagenser Kloningssæt og ligering
The original QuikChange Site-Directed Mutagenesis Kits speed up and simplify site-directed mutagenesis studies. The kits eliminate the need for sub-cloning into M13-based bacteriophage vectors and for ss-DNA rescue. This allows oligo-mediated introduction of site-specific mutations into virtually any double-stranded plasmid DNA. In addition, the XL version of the kit is specially designed for efficient mutagenesis of large (4 to 14 kb) or otherwise difficult-to mutagenise plasmid templates. The XL kit features components specifically designed for more efficient DNA replication and bacterial transformation.
  • >80% mutagenesis efficiency for one site; >55% mutagenesis efficiency for three mutations simultaneously
  • Efficient and accurate linear amplification minimises unwanted errors
QuikChange Site-Directed Mutagenesis Kit: This kit uses PfuTurbo DNA polymerase. It replicates plasmid strands with high fidelityll without displacing mutant oligonucleotide primers. The process uses a supercoiled dsDNA vector with insert and two synthetic oligo primers containing the mutation. Incorporating oligo primers creates a mutated plasmid with staggered nicks. The product is treated with Dpn I after temperature cycling. Dpn I digests the parental DNA template, selecting mutation-containing synthesised DNA. The nicked vector DNA containing the mutations is transformed into XL1-Blue supercompetent cells.
QuikChange XL Site-Directed Mutagenesis Kit: The QuikChange II XL site-directed mutagenesis kit specialises in efficient mutagenesis of large (8 to14 kb) or otherwise difficult-to mutagenise plasmid templates and features components designed for more efficient DNA replication and bacterial transformation. A QuikSolution reagent is provided to facilitate replication of large plasmids, while XL10-Gold Ultracompetent Cells have been included to ensure the highest transformation efficiencies possible. XL10-Gold cells contain the Hte phenotype, increasing the transformation efficiency of larger DNA plasmids.
QuikChange Multi site-directed mutagenesis kit: The QuikChange Multi site-directed mutagenesis system is a novel technology that allows mutagenesis at multiple sites in a single round, using a single oligonucleotide per site. This system simplifies randomising key amino acids using oligos containing degenerate codons. No specialised vectors or unique restriction sites are needed to use the kit; virtually any plasmid up to 8 kb is a suitable template. The rapid three-step procedure introduces mutations at three different sites simultaneously in the 4 kb QuikChange Multi control plasmid with >50% efficiency.
Advarsel: Academic QuikChange Multi site-directed mutagenesis Kits are for Academic Use Only.