ReadiUse™ Yeast Cell Lysis Buffer, 5X
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24102
AATB24102EA 1150 DKK
AATB24102
ReadiUse™ Yeast Cell Lysis Buffer, 5X
Buffere Ekstraktions buffere
ReadiUse™ Yeast Cell Lysis Buffer is a biochemical reagent designed for efficient yeast cell lysis and protein solubilisation in laboratory applications.
  • Designed for ease of use
  • Saves time
  • Versatile in its efficacy across various organisms, including Bacillus subtilis, Gram-positive, and Gram-negative bacteria
With a mild detergent formulation, ReadiUse™ Yeast Cell Lysis Buffer minimises the reliance on harsh chemical treatments, extreme pH adjustments, or temperature variations, contributing to the preservation of extracted protein integrity.
Advarsel: For research use only.