VWR® Xtra Nitrile Green Glove
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWRI112-6650EA 195 DKK
VWRI112-6650 VWRI112-6649 VWRI112-6648 VWRI112-6651 VWRI112-6647
VWR® Xtra Nitrile Green Glove
Handsker
These single-use, disposable examination protective gloves are made from nitrile and are developed with sustainability in mind. They are suitable for use in medical applications, food processing, and for general lab requirements in non-hazardous areas.
  • Non sterile, powder-free
  • Free from latex, chlorine, and accelarators
  • AQL 1.0
  • Food contact approved
The environmental impact of this glove has been assessed via Life Cycle Analysis meeting ISO 14040 and ISO 14044. Reduce your eco-footprint while still wearing a glove that is skin-friendly, and provides superior barrier protection against blood-borne pathogens and viruses.
Certificeringer:
EN ISO 374-1 Type B (JKT), EN 374-5 VIRUS, MDR Class I; PPE Cat. III; AQL 1.5, food contact approved.
Emballering:
Supplied 200 per bag (or 180 for size XL). 10 dispensers/carton. Less packaging waste.