Vios™ iDx CO₂ Incubators
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
51036156 51036162 51036153 51036159 51036276 51036282 51036273 51036279
390-1929EA 117630 DKK
390-1929 390-1930 390-1931 390-1932 390-1933 390-1934 390-1935 390-1936
Vios™ iDx CO₂ Incubators
Inkubatorer CO2-inkubatorer
Specifically designed to deliver the performance reliability, ease of operation and value required to support a range of culturing needs from basic research to demanding leading-edge applications. Specifically designed to deliver the performance reliability, ease of operation and value required to support a range of culturing needs from basic research to demanding leading-edge applications.
  • Thermo Scientific™ THRIVE™ Active Airflow technology for homogenous growth conditions
  • Thermo Scientific™ Steri-Run™ High Temperature Sterilisation Cycle delivers 12-log sterility assurance
  • In-chamber HEPA filtration to achieve ISO Class 5 cleanroom like air quality within only 5 minutes after a 30-second door opening
  • High quality in-chamber infrared sensor for CO2 (IR180Si or TC180), temperature, and optional O2 enables fast reactions to help maintain desired conditions
  • Large 18 cm (7”) touchscreen display for quicker at-a-glance monitoring of chamber conditions
  • Protected, integrated humidity reservoir helps keep an optimal environment and minimises contamination
  • Ergonomic door design to make routine loading, unloading, and checking on cultures easier
  • Sterilization cycle temperature: 180 °C
  • Sterilization cycle duration: ≤12 hrs
Invest in a Vios iDx CO2 Incubator and you can benefit not just from top-tier cell expansion performance, but from the confidence that comes with knowing you're ready to tackle whatever challenges tomorrow may bring.
Certificeringer:
UL and CSA compliant.
Bestillingsoplysninger:
Accessories are not included and must be ordered separately.