EZ-Fit® Family Kit
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KWEFHAW10B KWEFHAW25B KWEFHAW10S
MILXKWEFHAW10BEA 0 DKK
MILXKWEFHAW10B MILXKWEFHAW25B MILXKWEFHAW10S
EZ-Fit® Family Kit
Manifolder
EZ-Fit® filtration system for bioburden testing.
  • Sterility electron beam irradiation
  • Ready-to-use
  • Agency: ISO 7704
  • Depth: 47 mm
Materials are Mixed Cellulose Esters (MCE) membrane, polystyrene cover, stainless steel (manifold), styrene-butadiene funnel(s), white filter.
Emballering:
Pack of 48 units.