EZ-Pak® Membranes for use with EZ-Pak® Dispenser Curve
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EZAABG474 EZHCWG474 EZAAWG474
MILXEZAABG474EA 0 DKK
MILXEZAABG474 MILXEZHCWG474 MILXEZAAWG474
EZ-Pak® Membranes for use with EZ-Pak® Dispenser Curve
Filtre Membranfiltre
High throughput: Each cartridge contains 150 membranes. Less frequent reloading saves time.
Visible traceability: The membrane catalog number, lot number, pore size and sequential number (from 150 to 1) are printed on the protective film of every membrane cell.
  • From 0,2 to 0,8 μm pore size
  • Diameters of 47 mm
  • White, black and green
Certificeringer:
ISO® 7704:1985, ISO® 16266: 2006, ISO® 14189: 2013, ISO® 17995 : 2005, ISO® 8199:2005, ISO® 7899-2:2000, ISO® 9308-1: 2014, ISO® 19250: 2010, ISO® 6461-2:1986.