ESD Safety Shoes, Low, Red, IAN XXTP, S2
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
721310/35 721310/36 721310/37 721310/38 721310/39 721310/40 721310/41 721310/42 721310/43 721310/44 721310/45 721310/46 721310/47 721310/48 721310/49
ELTN721310/35EA 738 DKK
ELTN721310/35 ELTN721310/36 ELTN721310/37 ELTN721310/38 ELTN721310/39 ELTN721310/40 ELTN721310/41 ELTN721310/42 ELTN721310/43 ELTN721310/44 ELTN721310/45 ELTN721310/46 ELTN721310/47 ELTN721310/48 ELTN721310/49
ESD Safety Shoes, Low, Red, IAN XXTP, S2
Sko og støvler
Unisex IAN XXTP red Low ESD S2 safety shoes with excellent wearing comfort where the padded upper edge protects the Achilles tendon. The tongue prevents pressure marks and avoids dirt from entering into the shoe. The shoes gives  a good visibility in the dark.
  • Hydrophobised microfiber/hydrophobised textile material
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black with recycled material content
  • Soft fleece ESD insole
  • TPU/PU sole Wellmaxx Trainers Power
  • Composite toe cap and breathable fabric lining
  • Metal- and leather-free equipment, sole core made of Infinergy® by BASF
The sole core consists of expanded, thermoplastic polyurethane in the form of oval foam beads. These stick together and are very light and elastic. This revolutionary technology cushions the impact and bounces back extremely well on pressure, so that the energy can be returned to the wearer. Even under low temperatures of –20 °C, the core maintains its high elasticity.
Application: 
Indoors and outdoors
Areas where exposure to moisture is expected (S2)
Areas where there is a risk of electrostatic discharge (ESDS/ESD) E.g. airports, airplane construction, automobile manufacturing
No scratches from metal parts
Close to induction loops or metal detectors
Certificeringer:
EN ISO 20345:2022 S2 FO/SR/SC, form A.