Loggers with Bottle and Can Adapters for Pasteurization and Lab Use, SL412X Set: EBI 12-T46X
Leverandør: EBRO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1250-4121 1250-4122 1250-4123 1250-4124 1250-4125
EBRO1250-4121EA 58550 DKK
EBRO1250-4121 EBRO1250-4122 EBRO1250-4123 EBRO1250-4124 EBRO1250-4125
Loggers with Bottle and Can Adapters for Pasteurization and Lab Use, SL412X Set: EBI 12-T46X
Dataloggere
The SL412X set by ebro, featuring EBI 12-T46X loggers with the EBI FL-S bottle adapter and EBI DA can adapter, is ideal for precise temperature monitoring in pasteurization and laboratory applications, ensuring reliable data collection in both food and beverage production and lab research settings.
  • Versatile data logger set for pasteurization and lab applications
  • Includes EBI 12-T46X temperature loggers, EBI FL-S bottle adapter, and EBI DA can adapter
  • Suitable for temperature monitoring in bottles, cans, and lab samples
  • Traceablel factory calibration certificate
The SL412X set from ebro is a comprehensive solution for temperature validation in both industrial pasteurization and laboratory applications. Equipped with EBI 12-T46X temperature loggers, the set includes the EBI FL-S bottle adapter and EBI DA can adapter, enabling precise internal temperature monitoring in bottles, cans, and lab samples. The high-precision loggers are designed to support optimal process adjustments for quality control, energy efficiency, and consistent data collection. Suitable for rigorous lab environments, the SL412X set’s compact and IP-rated design ensures dependable performance, making it essential for maintaining quality standards in both food production and scientific research.
Dimensions: Ø 48×24H mm
Calibration points: –20, 0, +60 and 134 °C
Leveringsinformation:
EBI 12-T46× temperature data logger, 1× EBI FL-S bottle adapter, 1× EBI DA can adapter,  1× EBI IF 100 interface, 1x Winlog.pro software, 1× carrying case SYSTAINER.