Lab Chairs, Texon
Leverandør: bimos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9173-4000 9175-4000
132-1466EA 7350 DKK
132-1466 132-1467
Lab Chairs, Texon
Stole
The laboratory Texon chairs set new standards in the laboratory. With its consistent hygienic design and outstanding ergonomic concept, it is a leader among laboratory chairs. Thanks to the smooth, closed surfaces and the clean cover, under which the mechanism is hidden, cleaning is particularly easy and uncomplicated.
  • Intuitive and simple operation
  • Seat height adjustment
  • Auto-Motion technology for active dynamic sitting and optimum support in any sitting position (Can be fixed in three settings)
  • The Flex-Back on the lower backrest increases the range of movement to the rear
  • Automatic weight adjustment
  • Extra thick seat upholstery for outstanding seating comfort
  • Replaceable upholstery system allows cushions to be easily removed and replaced as required
  • Aluminium base (ø 700 mm), anthracite
  • Max. load capacity 150 kg
Hygienic design - Smooth, closed surfaces and seamless design. Clean cover; Covers the mechanism for particularly easy cleaning. Disinfectable, resistant to chemicals and easy to keep clean.
Skintec is the innovative upholstery that combines first-class seating comfort with exceptional ease of cleaning and durability The surface is resistant to all disinfectants. Skintec is manufactured in a particularly sustainable and environmentally friendly way.
Certificeringer:
Can be used in biotechnology laboratories in accordance with safety classes S1, S2, S3 of the Biological Agents Regulation. GMP-compliant.