Arctic Express™ Cryogenic Shippers with Datalogger
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CY50905D CY50910D
BARNCY50905DEA 38510 DKK
BARNCY50905D BARNCY50910D
Arctic Express™ Cryogenic Shippers with Datalogger
Dewars og kryogene fartøjer
Thermo Scientific™ Arctic Express™ cryogenic shippers are designed to ensure the maintenance of sample integrity when movement of your samples is required. Arctic Express shippers come available with a built-in 21CFR, part 11 compliant datalogger, providing you with additional peace of mind that your important samples have been maintained during transport.
  • Innovative material absorbs liquid nitrogen to prevent spillage during shipment
  • 14- and 21-day liquid nitrogen holding times allow specimens to be shipped safely
  • Durable construction and sturdy base allow dry shipper to withstand rough handling
  • Lockable lid prevents unauthorised entry
  • Easily transported via most common carriers
  • Lightweight aluminum design and convenient pail-style handle
Manual fill LN₂ Dewar type.
Advarsel:
Warranty varies by country, please contact us for details.