TriLink® N1-Methyl-Pseudouridine-5'-Triphosphate
Leverandør: TriLink BioTechnologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N-1081-10 N-1081-1 N-1081-100 N-1081-5
SOLUN-1081-10EA 3530 DKK
SOLUN-1081-10 SOLUN-1081-1 SOLUN-1081-100 SOLUN-1081-5
TriLink® N1-Methyl-Pseudouridine-5'-Triphosphate
Nukleinsyrereagenser Nukleotider
N1-Methyl-Pseudouridine-5'-Triphosphate is widely used in mRNA synthesis for therapeutic and vaccine development to improve mRNA functionality. This nucleotide consists of a modified base, a ribose, and a 5′ triphosphate group.
  • Reduces mRNA immunostimulatory effects
  • Improves mRNA translation efficiency
  • Enhances mRNA stability
  • ≥99% purity by AX-HPLC analysis
  • Provided as 100 mM in H₂O, pH 7,5
  • Grade: RUO
N1-Methylpseudouridine-5'-Triphosphate (N1MePsU) is a modified NTP for incorporation into messenger RNAs (mRNA) using an enzyme such as T7 RNA Polymerase. Modified uridines such as N1-methylpseudouridine, 5-methoxyuridine, and pseudouridine can reduce unwanted immunogenic response and enhance translational efficiency of mRNAs. These properties can result in safer mRNA and increased protein expression.
Advarsel: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80b8a387fb2b06b759d6fdc97bfd1d5ed8d90eba&vid=80b8a387fb2b06b759d6fdc97bfd1d5ed8d90eba&prid=web&clid=SAPDAM