TriLink® Sulf Cyanine5 Aminoallyluridine-5'-Triphosphate
Leverandør: TriLink BioTechnologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N-5110-0250 N-5110-1 N-5110-5
SOLUN-5110-0250EA 5120 DKK
SOLUN-5110-0250 SOLUN-5110-1 SOLUN-5110-5
TriLink® Sulf Cyanine5 Aminoallyluridine-5'-Triphosphate
Nukleinsyrereagenser Nukleotider
Cyanine 5-Aminoallyluridine-5'-Triphosphate is a substrate for T7 RNA polymerase, allowing the incorporation of cyanine 5 into RNA and mRNA during in vitro transcription. Cyanine 5 incorporation allows for fluorescent detection of RNA and mRNA independent of protein translation.
  • Grade: RUO
Advarsel: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.