TriLink® 5-Propargylamino-2'-deoxyuridine-5'-Triphosphate
Leverandør: TriLink BioTechnologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N-2062-1 N-2062-10 N-2062-5
SOLUN-2062-1EA 2380 DKK
SOLUN-2062-1 SOLUN-2062-10 SOLUN-2062-5
TriLink® 5-Propargylamino-2'-deoxyuridine-5'-Triphosphate
Nukleinsyrereagenser Nukleotider
5-Propargylamino-2'-deoxyuridine can be enzymatically incorporated into DNA and cDNA to introduce primary amine groups. Primary amines can be labeled with NHS ester fluors and haptens for detection and immobilisation of the resulting DNA strand.
  • Grade: RUO
Advarsel: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=671ff8f8b79325a9a8fb9644620de244939604b9&vid=671ff8f8b79325a9a8fb9644620de244939604b9&prid=web&clid=SAPDAM