Endeavor™ Orbital Shakers
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30770916 30770915
OHAU30770916EA 9780 DKK
OHAU30770916 OHAU30770915
Endeavor™ Orbital Shakers
Rystere Rysteapparater med cirkulære bevægelser
Stable, long-term shaking with triple eccentric drive and maintenance-free brushless DC motor.
  • Adaptable to a wide variety of accessories to customize for low to mid volume applications
  • Programmable with a simple on-board menu to save time and guarantee reproducible results
  • Remote function permits PC control and data transfer
For use in a wide variety of applications including microbiology, molecular biology, cell cultures, staining/destaining, DNA studies, environmental and food sciences, chemistry, general mixing and more.
Feature a bright, backlit LCD display for speed and time. The timer will count up elapsed time or when set, timer will count down and shut off shaking when time reaches zero. A green indicator light illuminates when the unit is shaking with visual and audio alarm when timer counts down to zero or an error occurs.
Small footprint to save bench space, durable chemical-resistant polymer housing, simple knob control for timer and shaking, easy to navigate menu for advanced features.
Certificeringer:
Product Safety: IEC/EN 61010-1; IEC/EN 61010-2-051, Electromagnetic Compatibility: IEC/EN 61326-1 Class B, Basic electromagnetic environment, Compliance Marks:  TÜV SÜD, IP protection class according to DIN EN 60529: IP 21.
Leveringsinformation:
Supplied with a sturdy, easy-to-clean aluminum tray with attachment points for flask clamps, tube racks and non-slip rubber mat. Optional accessories include universal platform and over 20 flask clamps and tube racks.