Human IL-10 ELISA Kit
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB46034-96 AB46034-2X96
ABCAAB46034-96EA 7230 DKK
ABCAAB46034-96 ABCAAB46034-2X96
Human IL-10 ELISA Kit
Assays ELISA'er
Human IL-10 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-10 in Human in plasma, cell culture supernatant, serum samples.