TMB ELISA Substrate (High Sensitivity)
Leverandør: Abcam
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB171523-100
AB171523-1000
ABCAAB171523-100EA
746
DKK
ABCAAB171523-100
ABCAAB171523-1000
TMB ELISA Substrate (High Sensitivity)
Assays
ELISA'er
Abcam's TMB ELISA Substrate (High Sensitive) detects horseradish peroxidase (HRP) activity and contains 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer.