phynomic lite Safety Gloves, 60040 Series
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6004005 6004012
UVEX6004005EA 455 DKK
UVEX6004005 UVEX6004012
phynomic lite Safety Gloves, 60040 Series
Handsker
The phynomic series is ideal for all types of work where a high level of feeling is required. These safety gloves are like a 'second skin' and are especially light and flexible.
  • Exceptionally light and flexible
  • Good grip in dry and slightly damp areas
  • Food-safe
  • Excellent skin tolerance and free from pollutants
The uvex phynomic lite is extremely lightweight, with a composition that makes it suited to particularly delicate tasks. They are free of accelerators and dermatologically tested for skin compatibility, making them ideal for allergy sufferers. These gloves also offer a very high level of breathability with the porous coating, which reduces sweating.