u-chem 3500 Chemical Protection Gloves, 60188 Series
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6018810 6018811 6018807 6018808
UVEX6018810EA 1360 DKK
UVEX6018810 UVEX6018811 UVEX6018807 UVEX6018808
u-chem 3500 Chemical Protection Gloves, 60188 Series
Handsker
The uvex u-chem 3500 chemical protection gloves are ideal for handling a variety of different chemicals.
  • Very comfortable to wear thanks to seamless cotton liner
  • Fully coated with chloroprene and nitrile rubber (NBR)
  • Excellent grip on warm and wet workpieces
  • High resistance to acetone, cleaning agents, adhesives and solvents
Thanks to their full-surface chloroprene and NBR coating, the gloves offer excellent grip and are resistant to acetone, cleaning agents, adhesives, and solvents. These gloves are very comfortable to wear thanks to a seamless cotton liner.