Anti-FGFR2IIIb/FGFR2b Rabbit Monoclonal Antibody [clone: EPR28355-27]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB318274-20UL AB318274-100UL AB318274-1ML
ABCAAB318274-20ULEA 1820 DKK
ABCAAB318274-20UL ABCAAB318274-100UL ABCAAB318274-1ML
Anti-FGFR2IIIb/FGFR2b Rabbit Monoclonal Antibody [clone: EPR28355-27]
Antistoffer
Rabbit monoclonal [EPR28355-27] to FGFR2IIIb/FGFR2b.
Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FGFR2IIIb/FGFR2b
Clonality: Monoclonal
Clone: EPR28355-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human