Anti-E2F2 Rabbit Monoclonal Antibody [clone: EPR27391-170]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB318956-20UL AB318956-100UL AB318956-1ML
ABCAAB318956-20ULEA 1820 DKK
ABCAAB318956-20UL ABCAAB318956-100UL ABCAAB318956-1ML
Anti-E2F2 Rabbit Monoclonal Antibody [clone: EPR27391-170]
Antistoffer
Rabbit monoclonal [EPR27391-170] to E2F2.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: E2F2
Clonality: Monoclonal
Clone: EPR27391-170
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR