Anti-ITGA11 Rabbit Monoclonal Antibody [clone: EPR28689-21]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB316249-20UL AB316249-100UL AB316249-1ML
ABCAAB316249-20ULEA 1820 DKK
ABCAAB316249-20UL ABCAAB316249-100UL ABCAAB316249-1ML
Anti-ITGA11 Rabbit Monoclonal Antibody [clone: EPR28689-21]
Antistoffer
Rabbit monoclonal [EPR28689-21] to ITGA11.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ITGA11
Clonality: Monoclonal
Clone: EPR28689-21
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR