Anti-HLA F Mouse Monoclonal Antibody [clone: 3D11]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB314768-1MG AB314768-100UG
ABCAAB314768-1MGEA 68310 DKK
ABCAAB314768-1MG ABCAAB314768-100UG
Anti-HLA F Mouse Monoclonal Antibody [clone: 3D11]
Antistoffer
Mouse monoclonal [3D11] to HLA F - BSA and Azide free.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HLA F
Clonality: Monoclonal
Clone: 3D11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human