Anti-BLNK (phospho Y84) Rabbit Monoclonal Antibody [clone: BLNKY84-H4] (APC (Allophycocyanin))
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB278596-100 AB278596-10
ABCAAB278596-100EA 6110 DKK
ABCAAB278596-100 ABCAAB278596-10
Anti-BLNK (phospho Y84) Rabbit Monoclonal Antibody [clone: BLNKY84-H4] (APC (Allophycocyanin))
Antistoffer
APC Rabbit monoclonal [BLNKY84-H4] to BLNK (phospho Y84).
Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: BLNK
Clonality: Monoclonal
Clone: BLNKY84-H4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human