Anti-TDP43 Mouse Monoclonal Antibody [clone: DB9]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB255922-100 AB255922-1
ABCAAB255922-100EA 7600 DKK
ABCAAB255922-100 ABCAAB255922-1
Anti-TDP43 Mouse Monoclonal Antibody [clone: DB9]
Antistoffer
Mouse monoclonal [DB9] to TDP43 - BSA and Azide free.

Type: Primary
Antigen: TDP43
Clonality: Monoclonal
Clone: DB9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR