Anti-PGAM5 Rabbit Monoclonal Antibody [clone: EPR27145-82]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB308449-100 AB308449-20UL AB308449-1ML
ABCAAB308449-100EA 5180 DKK
ABCAAB308449-100 ABCAAB308449-20UL ABCAAB308449-1ML
Anti-PGAM5 Rabbit Monoclonal Antibody [clone: EPR27145-82]
Antistoffer
Rabbit monoclonal [EPR27145-82] to PGAM5.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PGAM5
Clonality: Monoclonal
Clone: EPR27145-82
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human