Anti-ADA Rabbit Monoclonal Antibody [clone: EPR25429-117]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB300050-100 AB300050-20UL AB300050-1ML
ABCAAB300050-100EA 5270 DKK
ABCAAB300050-100 ABCAAB300050-20UL ABCAAB300050-1ML
Anti-ADA Rabbit Monoclonal Antibody [clone: EPR25429-117]
Antistoffer
Rabbit monoclonal [EPR25429-117] to ADA.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ADA
Clonality: Monoclonal
Clone: EPR25429-117
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR