Anti-LAG-3 Rabbit Monoclonal Antibody [clone: EPR4392(2)]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB209740-100 AB209740-1
ABCAAB209740-100EA 7600 DKK
ABCAAB209740-100 ABCAAB209740-1
Anti-LAG-3 Rabbit Monoclonal Antibody [clone: EPR4392(2)]
Antistoffer
Rabbit monoclonal [EPR4392(2)] to LAG-3 - BSA and Azide free.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LAG-3
Clonality: Monoclonal
Clone: EPR4392(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR