Anti-AWP1 Rabbit Monoclonal Antibody [clone: EPR12323]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB180856-100 AB180856-1ML
ABCAAB180856-100EA 5090 DKK
ABCAAB180856-100 ABCAAB180856-1ML
Anti-AWP1 Rabbit Monoclonal Antibody [clone: EPR12323]
Antistoffer
Rabbit monoclonal [EPR12323] to AWP1.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: AWP1
Clonality: Monoclonal
Clone: EPR12323
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR