Anti-CCR2 Rabbit Monoclonal Antibody [clone: EPR23494-47] (APC (Allophycocyanin))
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB313950-10UL AB313950-100UL
ABCAAB313950-10ULEA 1780 DKK
ABCAAB313950-10UL ABCAAB313950-100UL
Anti-CCR2 Rabbit Monoclonal Antibody [clone: EPR23494-47] (APC (Allophycocyanin))
Antistoffer
APC Rabbit monoclonal [EPR23494-47] to CCR2.
Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: EPR23494-47
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human