Anti-Bassoon/BSN Mouse Monoclonal Antibody [clone: L124-59]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB306593-100 AB306593-10 AB306593-20UG AB306593-1MG
ABCAAB306593-100EA 5180 DKK
ABCAAB306593-100 ABCAAB306593-10 ABCAAB306593-20UG ABCAAB306593-1MG
Anti-Bassoon/BSN Mouse Monoclonal Antibody [clone: L124-59]
Antistoffer
Mouse monoclonal [L124-59] to Bassoon/BSN.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Bassoon/BSN
Clonality: Monoclonal
Clone: L124-59
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR