Anti-CCR2 Rabbit Monoclonal Antibody [clone: EPR23494-47] (PE (Phycoerythrin))
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB314292-100UL AB314292-10UL
ABCAAB314292-100ULEA 6110 DKK
ABCAAB314292-100UL ABCAAB314292-10UL
Anti-CCR2 Rabbit Monoclonal Antibody [clone: EPR23494-47] (PE (Phycoerythrin))
Antistoffer
PE Rabbit monoclonal [EPR23494-47] to CCR2.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: EPR23494-47
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human